Please login to your online account to display your discounted pricing

Lithium foil, 99.9% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Lithium foil
CAS 7439-93-2
Assay Percent Range 99.9% (metals basis)
MDL Number MFCD00134051
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1076722

 
alfa aesar
1076722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material Lithium foil
EINECS Number 231-102-5
Assay Percent Range 99.9% (metals basis)
MDL Number MFCD00134051
CAS 7439-93-2