missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium fluoride, White powder, 99.99% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1446309

 View more versions of this product

Catalog No. AA1446309



Specifications

Specifications

Lithium fluoride
99.99% (Metals basis)
MFCD00011090
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium;fluoride
224478
Powder
Odorless
10g
7789-24-4
FLi
lithium fluoride, fluorolithium, lithium fluoride, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
[Li+].[F-]
25.938
25.94
2.635g/cm3 at 20°C
White

Chemical Identifiers

7789-24-4
25.938
PQXKHYXIUOZZFA-UHFFFAOYSA-M
224478
[Li+].[F-]
FLi
MFCD00011090
lithium fluoride, fluorolithium, lithium fluoride, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
lithium;fluoride
Safety and Handling

Safety and Handling

Keep away from living quarters. Keep container in a well ventilated place. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H301-H315-H319-H335: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 232-152-0

Documents