Please login to your online account to display your discounted pricing

Lithium fluoride, White powder, 99.99% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Lithium fluoride
CAS 7789-24-4
Physical Form Powder
Assay Percent Range 99.99% (Metals basis)
Molecular Formula FLi
Formula Weight 25.94
MDL Number MFCD00011090
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1446309

 
alfa aesar
1446309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1446318

 
alfa aesar
1446318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 25.94
Quantity 10g
Color White
DOT Information DOT Class: 6.1, Packaging Group: III
Odor Odorless
Chemical Name or Material Lithium fluoride
EINECS Number 232-152-0
Assay Percent Range 99.99% (Metals basis)
Molecular Formula FLi
MDL Number MFCD00011090
CAS 7789-24-4
Health Hazard 1 H301-H315-H319-H335: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 2 Keep away from living quarters. Keep container in a well ventilated place. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Density 2.635g/cm3 at 20°C
Safety and Handling

H301-H315-H319-H335: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

Keep away from living quarters. Keep container in a well ventilated place. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.