missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium fluoride, 99.99% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1073630

 View more versions of this product

Catalog No. AA1073630



Specifications

Specifications

Lithium fluoride
99.99% (Metal basis)
MFCD00011090
fluorolithium
[Li+].[F-]
25.938
25.94
Odorless
250g
7789-24-4
FLi
White powder
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium;fluoride
224478
Powder
White

Chemical Identifiers

7789-24-4
25.938
PQXKHYXIUOZZFA-UHFFFAOYSA-M
224478
[Li+].[F-]
FLi
MFCD00011090
fluorolithium
lithium;fluoride
Safety and Handling

Safety and Handling

H301-H315-H319-H335: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 232-152-0

Documents