missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium fluoride, 99.98% (metals basis)

Manufacturer:  Alfa Aesar™ 1098614

 View more versions of this product

Catalog No. AA1098614



Specifications

Specifications

Lithium fluoride
(metals basis)
LiF
UN3288
fluorolithium
[Li+].[F-]
25.938
25.94
99.98%
Odorless
1680°C
845°C
7789-24-4
FLi
MFCD00011090
14,5531
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium;fluoride
224478
Powder
2.635g/mL
1.3915
White
25g

Chemical Identifiers

7789-24-4
25.938
PQXKHYXIUOZZFA-UHFFFAOYSA-M
224478
[Li+].[F-]
FLi
MFCD00011090
fluorolithium
lithium;fluoride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : OJ6125000

TSCA : TSCA

Documents