missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium fluoride, 98.5%, Alfa Aesar™

For Use in Vitreous Enamels and Glazes, As Flux in Soldering and Welding Aluminum, Infrared Spectrophotometers

Manufacturer:  Alfa Aesar 1340618

 View more versions of this product

Catalog No. AA1340618



Specifications

Specifications

Lithium fluoride
7789-24-4
FLi
White powder
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium;fluoride
224478
Powder
White
Volatizes at 1100°-1200°
98.5%
MFCD00011090
lithium fluoride, fluorolithium, lithium fluoride, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
[Li+].[F-]
25.938
25.94
Odorless
50g

Chemical Identifiers

7789-24-4
25.938
PQXKHYXIUOZZFA-UHFFFAOYSA-M
224478
[Li+].[F-]
FLi
MFCD00011090
lithium fluoride, fluorolithium, lithium fluoride, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
lithium;fluoride
Safety and Handling

Safety and Handling

H301-H315-H319-H335: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 232-152-0

Documents