Please login to your online account to display your discounted pricing

Lithium fluoride, 97%, for analysis, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  218271000

 View more versions of this product

Catalog No. AC218271000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 25.93
Quantity 100g
Color White to off-white
Purity Grade For analysis
Density 2.63g/mL
Calcium (Ca) 0.08% max.
Carbonate 1.0% max.
Sodium (Na) 0.4% max.
CAS 7789-24-4
Magnesium (Mg) 0.02% max.
Phosphate 0.04% max.
Chemical Name or Material Lithium fluoride
Assay Percent Range 97%
Trace Analysis Trace analysis are typical values
Loss on Drying 0.4% (200°C) (typical)
Packaging Plastic bottle
Potassium (K) 0.07% max.
Chlorine (Cl) 0.07% max.
Iron (Fe) 0.03% max.
Sulfate 1.0% max.
Linear Formula LiF
Molecular Formula FLi

Specifications

Chemical Name or Material Lithium fluoride
CAS 7789-24-4
Purity Grade For analysis
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula FLi
Linear Formula LiF
Formula Weight 25.93