Please login to your online account to display your discounted pricing

Lithium ethoxide, 1M solution in THF, Acros Organics


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC380181000

 View more versions of this product

Catalog No. AC380181000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Harmful if swallowed. May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Formula Weight 52
Quantity 100mL
Packaging AcroSeal™ Glass Bottle
Concentration 1M, exact strength on the certificate of analysis
Density 0.8950g/mL
Appearance of Solution brown solution
CAS 109-99-9
CAS 2388-07-0
Molecular Formula C2H5LiO
MDL Number MFCD00050493
Name Note 1M solution in THF
Chemical Name or Material Lithium ethoxide
Linear Formula LiOCH2CH3
Health Hazard 1 Danger

Specifications

Chemical Name or Material Lithium ethoxide
Name Note 1M solution in THF
CAS 2388-07-0
CAS 109-99-9
Molecular Formula C2H5LiO
Linear Formula LiOCH2CH3
Formula Weight 52
MDL Number MFCD00050493
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Harmful if swallowed. May cause drowsiness or dizziness.

SDS & Safety Documents