Please login to your online account to display your discounted pricing

Lithium ethoxide, 1M solution in THF, Acros Organics

Chemical Identifiers

Chemical Name or Material Lithium ethoxide
Name Note 1M solution in THF
CAS 2388-07-0
CAS 109-99-9
Molecular Formula C2H5LiO
Linear Formula LiOCH2CH3
Formula Weight 52
MDL Number MFCD00050493
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380181000

  SDS
thermo fisher scientific
AC380181000
AcroSeal™ Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 52
Quantity 100mL
Packaging AcroSeal™ Glass Bottle
Concentration 1M, exact strength on the certificate of analysis
Density 0.8950g/mL
Appearance of Solution brown solution
CAS 109-99-9
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for se
CAS 2388-07-0
Molecular Formula C2H5LiO
MDL Number MFCD00050493
Name Note 1M solution in THF
Chemical Name or Material Lithium ethoxide
Linear Formula LiOCH2CH3
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water.
Safety and Handling

Danger

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for se

GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water.