Please login to your online account to display your discounted pricing

Lithium ethoxide, 1M solution in ethanol, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC380178000

 View more versions of this product

Catalog No. AC380178000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 52
Quantity 800mL
Color Clear orange
Packaging AcroSeal™ Glass Bottle
Density 0.8200g/mL
Identification Confirmed
CAS 64-17-5
Linear Formula LiOCH2CH3
Concentration Exact strength on the certificate of analysis
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Lithium ethoxide
MDL Number MFCD00050493
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 2388-07-0
Molecular Formula C2H5LiO
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Reacts violently with water.
Name Note 1M solution in ethanol

Specifications

Chemical Name or Material Lithium ethoxide
Name Note 1M solution in ethanol
CAS 2388-07-0
CAS 64-17-5
Molecular Formula C2H5LiO
Linear Formula LiOCH2CH3
Formula Weight 52
MDL Number MFCD00050493
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Reacts violently with water.

SDS & Safety Documents