Please login to your online account to display your discounted pricing

Lithium ethoxide, 1M solution in ethanol, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC380171000

 View more versions of this product

Catalog No. AC380171000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 52
Quantity 100mL
Color Clear orange
Packaging AcroSeal™ Glass Bottle
Density 0.8200g/mL
Identification Confirmed
Concentration Exact strength on the certificate of analysis
MDL Number MFCD00050493
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Reacts violently with water.
Linear Formula LiOCH2CH3
Chemical Name or Material Lithium ethoxide
CAS 64-17-5
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C2H5LiO
CAS 2388-07-0
Name Note 1M solution in ethanol
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Lithium ethoxide
Name Note 1M solution in ethanol
CAS 2388-07-0
CAS 64-17-5
Molecular Formula C2H5LiO
Linear Formula LiOCH2CH3
Formula Weight 52
MDL Number MFCD00050493
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Reacts violently with water.

SDS & Safety Documents