missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium ethoxide, 1M solution in ethanol, ACROS Organics™

Chemical Identifiers

CAS 2388-07-0,64-17-5
Molecular Formula C2H5LiO
Linear Formula LiOCH2CH3
Formula Weight 52
MDL Number MFCD00050493
Chemical Name or Material Lithium ethoxide, 1M solution in ethanol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC380171000 SDS Thermo Fisher Scientific
AC380171000
AcroSeal™ Glass Bottle 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC380178000 SDS Thermo Fisher Scientific
AC380178000
AcroSeal™ Glass Bottle 800mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2388-07-0,64-17-5
LiOCH2CH3
MFCD00050493
C2H5LiO
52
Lithium ethoxide, 1M solution in ethanol

Specifications

2388-07-0,64-17-5
C2H5LiO
LiOCH2CH3
0.8200g/mL
Confirmed
Clear orange
Lithium ethoxide, 1M solution in ethanol
Liquid
Exact strength on the certificate of analysis
52
MFCD00050493
AcroSeal™ Glass Bottle
100mL

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Reacts violently with water.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger