Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium dodecylsulfate, 99+%

Chemical Identifiers

Chemical Name or Material Lithium dodecylsulfate
CAS 2044-56-6
Formula Weight 272.33
MDL Number MFCD00007467
UN Number UN1325
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3932809

  SDS
alfa aesar™
3932809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3932818

  SDS
alfa aesar™
3932818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 272.33
Chemical Name or Material Lithium dodecylsulfate
TSCA TSCA
Physical Form Powder
Assay ≥99%
MDL Number MFCD00007467
CAS 2044-56-6
Quantity 10g
UN Number UN1325
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H228-H335-H315
Causes serious eye damage.
Flammable solid.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White
Odor Odorless
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

  • Detergent for solubilizing proteins for electrophoresis which exhibits greater solubility than SDS at lower temperatures

Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H228-H335-H315
Causes serious eye damage.
Flammable solid.
May cause respiratory irritation.
Causes skin irritation.