missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Dodecyl Sulfate (Powder or Flakes/Molecular Biology), Fisher BioReagents

$101.66 - $101.66

Chemical Identifiers

CAS 2044-56-6
Molecular Formula C12H25LiO4S
Molecular Weight (g/mol) 272.33
MDL Number MFCD00007467
InChI Key YFVGRULMIQXYNE-UHFFFAOYSA-M
PubChem CID 2735071
IUPAC Name lithium(1+) dodecyl sulfate
SMILES [Li+].CCCCCCCCCCCCOS([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP1309-25
SDS View Documents Promotion Details
Fisher BioReagents
BP130925
25g Glass Bottle
Each for $101.66
 
Description

Description

Lithium Dodecyl Sulfate (LDS) is a detergent used for dissociating proteins.
Specifications

Chemical Identifiers

2044-56-6
272.33
YFVGRULMIQXYNE-UHFFFAOYSA-M
lithium(1+) dodecyl sulfate
C12H25LiO4S
MFCD00007467
2735071
[Li+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

0.01 max. (3% solution) at 405nm
≥99 %
C12H25LiO4S
Pass Test
Glass Bottle
1ppm max.
lithium(1+) dodecyl sulfate
2735071
Solid
Molecular Biology
25g
2044-56-6
5ppm max.
CH3(CH2)11OSO3Li
MFCD00007467
YFVGRULMIQXYNE-UHFFFAOYSA-M
[Li+].CCCCCCCCCCCCOS([O-])(=O)=O
272.33
273.18
≥99% (≥98% C12-OH)
White
Lithium Dodecyl Sulfate
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Product Certifications