Please login to your online account to display your discounted pricing

Lithium Dodecyl Sulfate (Powder or Flakes/Molecular Biology), Fisher BioReagents


Quantity 5g
Packaging Glass Bottle

Manufacturer: fisher scientific company  B13095

 View more versions of this product

Catalog No. BP1309-5

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99% (≥98% C12-OH)
Purity Grade Molecular Biology Grade
Formula Weight 273.18
Quantity 5g
Physical Form Solid
Molecular Formula C12H25LiO4S
Viscosity [NULL]
Color White
Absorbance 0.01 max. (3% solution) at 405nm
Identification Pass Test
Chemical Name or Material Lithium Dodecyl Sulfate
Heavy Metals (as Pb) 5ppm max.
Assay Percent Range ≥99 %
Linear Formula CH3(CH2)11OSO3Li
CAS 2044-56-6
ChemAlert Storage Symbol Gray
Packaging Glass Bottle
Phosphate 1ppm max.
Recommended Storage RT

Specifications

Chemical Name or Material Lithium Dodecyl Sulfate
CAS 2044-56-6
Purity Grade Molecular Biology Grade
Physical Form Solid
Assay ≥99% (≥98% C12-OH)
Assay Percent Range ≥99 %
Molecular Formula C12H25LiO4S
Linear Formula CH3(CH2)11OSO3Li
Formula Weight 273.18
Accessories
 

RT

SDS & Safety Documents