missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Dodecyl Sulfate [for Biochemical Research], TCI America™

Manufacturer:  TCI America L025425G

 View more versions of this product

Catalog No. L025425G



Specifications

Specifications

Lithium Dodecyl Sulfate [for Biochemical Research]
25g
C12H25LiO4S
lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure
[Li+].CCCCCCCCCCCCOS(=O)(=O)[O-]
272.328
272.33
≥98.0% (T)
White
2044-56-6
MFCD00007467
YFVGRULMIQXYNE-UHFFFAOYSA-M
lithium;dodecyl sulfate
2735071
Crystal-Powder at 20°C

Chemical Identifiers

2044-56-6
272.328
YFVGRULMIQXYNE-UHFFFAOYSA-M
2735071
[Li+].CCCCCCCCCCCCOS(=O)(=O)[O-]
C12H25LiO4S
MFCD00007467
lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure
lithium;dodecyl sulfate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3045&(8)-0606

TSCA : Yes

Documents