missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium dimethylamide, 96%

Manufacturer:  Alfa Aesar™ 4414506

 View more versions of this product

Catalog No. AA4414506



Specifications

Specifications

Lithium dimethylamide
Air and moisture sensitive
(decomposition)
3585-33-9
LiN(CH3)2
UN3393
acmc-20akms
YDGSUPBDGKOGQT-UHFFFAOYSA-N
lithium;dimethylazanide
5152226
Powder/Lump
Amine-like
White
5g
C2H6LiN
MFCD00008295
3618233
Reacts violently with water.
[Li+].C[N-]C
51.017
51.02
96%

Chemical Identifiers

3585-33-9
51.017
YDGSUPBDGKOGQT-UHFFFAOYSA-N
5152226
[Li+].C[N-]C
C2H6LiN
MFCD00008295
acmc-20akms
lithium;dimethylazanide
Safety and Handling

Safety and Handling

GHS H Statement
H250-H261-H314-H318
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P222-P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H250-H261-H314-H335

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC, WATER-REACTIVE

EINECSNumber : 222-714-3

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents