missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium diisopropylamide mono(tetrahydrofuran), 1.5M solution in cyclohexane, AcroSeal™, ACROS Organics™

Manufacturer:  Acros Organics AC432871000

 View more versions of this product

Catalog No. AC432871000



Specifications

Specifications

Lithium diisopropylamide mono(tetrahydrofuran)
product may develop some turbidity
lithium diisopropylamide mono tetrahydrofuran, lithium diisopropylamide mono tetrahydrofuran complex solution, lithium;di propan-2-yl azanide;oxolane, acmc-20ehmk, lda thf, lda.thf, lithiumdiisopropylamide thf, lithium diisopropylamide thf, lithiumdiisopropylamide tetrahydrofuran, lithium diisopropylamide tetrahydorfuran
[Li+].CC(C)[N-]C(C)C.C1CCOC1
179.232
179.23
1.43 to 1.65M
AcroSeal™ Glass Bottle
Brown to Orange-Yellow
−18°C
123333-84-6,110-82-7
C10H22LiNO
YTJXGDYAEOTOCG-UHFFFAOYSA-N
lithium;di(propan-2-yl)azanide;oxolane
10997623
Liquid
0.8160g/mL
0.816
100mL

Chemical Identifiers

123333-84-6
179.232
lithium diisopropylamide mono tetrahydrofuran, lithium diisopropylamide mono tetrahydrofuran complex solution, lithium;di propan-2-yl azanide;oxolane, acmc-20ehmk, lda thf, lda.thf, lithiumdiisopropylamide thf, lithium diisopropylamide thf, lithiumdiisopropylamide tetrahydrofuran, lithium diisopropylamide tetrahydorfuran
lithium;di(propan-2-yl)azanide;oxolane
C10H22LiNO
YTJXGDYAEOTOCG-UHFFFAOYSA-N
10997623
[Li+].CC(C)[N-]C(C)C.C1CCOC1
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents