Please login to your online account to display your discounted pricing

Lithium diisopropylamide mono(tetrahydrofuran), 1.5M solution in cyclohexane, AcroSeal™, ACROS Organics™

$56.52 - $279.41

Chemical Identifiers

Chemical Name or Material Lithium diisopropylamide mono(tetrahydrofuran)
CAS 123333-84-6
CAS 110-82-7
Molecular Formula C10H22LiNO
Formula Weight 179.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC432871000

  SDS
thermo fisher scientific
AC432871000
AcroSeal™ Glass Bottle 100mL Each for $56.52

AC432878000

  SDS
thermo fisher scientific
AC432878000
AcroSeal™ Glass Bottle 800mL Each for $279.41
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 179.23
Quantity 100mL
Color Yellow-orange to dark brown
Packaging AcroSeal™ Glass Bottle
Chemical Name or Material Lithium diisopropylamide mono(tetrahydrofuran)
Molecular Formula C10H22LiNO
CAS 123333-84-6
Health Hazard 1 Danger
Additional Information product may develop some turbidity
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
CAS 110-82-7
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Concentration 1.43 to 1.65M
Density 0.8160g/mL
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.