missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium diisopropylamide, 2M sol. in THF/n-heptane/ethylbenzene, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 268838000

 View more versions of this product

Catalog No. AC268838000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lithium diisopropylamide
2M Solution in THF/n-Heptane
60.0°C to 136.0°C
MFCD00064449
Lithium diisopropylamide: 26.0% (2.0M)
0.8100g/mL
lithium diisopropylamide, unii-ol028kiw1i, lithium di-isopropylamide, lithium diisopropyl amide, 2-propanamine, n-1-methylethyl-, lithium salt, ol028kiw1i, lithium di propan-2-yl azanide, 2-propanamine, n-1-methylethyl-, lithium salt 1:1, lithium n,n-diisopropylamide, ipr2nli
ZCSHNCUQKCANBX-UHFFFAOYSA-N
lithium;di(propan-2-yl)azanide
2724682
24.6 to 30.0% (Lithium diisopropylamide)
0.81
2°C
Brown-Red or Orange to Yellow
4111-54-0,142-82-5,109-99-9,100-41-4
C6H14LiN
Solvent: THF 59%, n-Heptane: 28%, Ethylbenzene: 13%
02,49; 03,184; 04,298; 05,400; 06,334; 07,204; 08,292; 09,280; 10,241; 11,296; 12,277; 14,193; 15,188; 16,15; 17,165
15, 3215
Solubility in water: decomposes. Other solubilities: soluble in thf, n-heptane, ethylbenzene, miscible with liquid aliphatic and aromatic, hydrocarbons, ethers and tertiary amines
[Li+].CC(C)[N-]C(C)C
107.125
107.12
AcroSeal™ Glass Bottle
800 mL
Solution

Chemical Identifiers

4111-54-0
107.125
ZCSHNCUQKCANBX-UHFFFAOYSA-N
2724682
[Li+].CC(C)[N-]C(C)C
C6H14LiN
MFCD00064449
lithium diisopropylamide, unii-ol028kiw1i, lithium di-isopropylamide, lithium diisopropyl amide, 2-propanamine, n-1-methylethyl-, lithium salt, ol028kiw1i, lithium di propan-2-yl azanide, 2-propanamine, n-1-methylethyl-, lithium salt 1:1, lithium n,n-diisopropylamide, ipr2nli
lithium;di(propan-2-yl)azanide
Safety and Handling

Safety and Handling

GHS P Statement May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. Toxic to aquatic life with long lasting effects. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

EINECSNumber : 223-893-0

TSCA : TSCA

Recommended Storage : Keep the solution at < 10°C at which it is stable

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only