Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium dihydrogen phosphate, 97%

$105.92 - $105.92

Chemical Identifiers

Chemical Name or Material Lithium dihydrogen phosphate
CAS 13453-80-0
Molecular Formula LiH2PO4
Formula Weight 103.93
MDL Number MFCD00016188
Synonym Lithium phosphate, monobasic; Phosphoric acid monolithium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1698730

  SDS
alfa aesar™
A1698730
250g Each for $105.92
Description & Specifications

Specifications

Formula Weight 103.93
Molecular Formula LiH2PO4
Health Hazard 1 Danger
Melting Point >100°C
MDL Number MFCD00016188
Odor Odorless
CAS 13453-80-0
Synonym Lithium phosphate, monobasic; Phosphoric acid monolithium salt
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 250g
Chemical Name or Material Lithium dihydrogen phosphate
TSCA TSCA

Applications
Employed as a precursor for LiMX2O7 (M = Fe, V; x =P, As) cathode materials.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.