missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium deuteride, 98%(Isotopic), Alfa Aesar™

Used in thermonuclear Fusion

Manufacturer:  Alfa Aesar 44100SS

 View more versions of this product

Catalog No. AA44100SS



Specifications

Specifications

Lithium deuteride
98% (Isotopic)
MFCD00011091
lithium deuteride, lithium hydride lid, 2h lithium, lithium deuteride 98 atom % d, lithium deuteride, 98 atom % d
[H-].[Li+]
8.954
8.95
200°C (392°F)
Gray
13587-16-1
HLi
Gray powder
SRTHRWZAMDZJOS-IEOVAKBOSA-N
lithium;deuteride
6914554
Powder
Odorless
0.2g

Chemical Identifiers

13587-16-1
8.954
SRTHRWZAMDZJOS-IEOVAKBOSA-N
6914554
[H-].[Li+]
HLi
MFCD00011091
lithium deuteride, lithium hydride lid, 2h lithium, lithium deuteride 98 atom % d, lithium deuteride, 98 atom % d
lithium;deuteride
Safety and Handling

Safety and Handling

H260-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 237-018-5

SDS Documents