Please login to your online account to display your discounted pricing

Lithium deuteride, 98%(Isotopic), Alfa Aesar™

Used in thermonuclear Fusion

Chemical Identifiers

Chemical Name or Material Lithium deuteride
CAS 13587-16-1
Physical Form Powder
Assay Percent Range 98% (Isotopic)
Molecular Formula DLi
Formula Weight 8.95
MDL Number MFCD00011091
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4410004

  SDS
alfa aesar
4410004
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA44100SS

  SDS
alfa aesar
44100SS
0.2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 8.95
Quantity 2g
Color Gray
DOT Information DOT Class: 4.3, Packaging Group: I
EINECS Number 237-018-5
MDL Number MFCD00011091
Density 0.906g/cm3 at 20°C
Appearance Gray powder
CAS 13587-16-1
Odor Odorless
Chemical Name or Material Lithium deuteride
Molecular Formula DLi
Assay Percent Range 98% (Isotopic)
Ignition Point 200°C (392°F)
Health Hazard 1 H260-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage.
Safety and Handling

H260-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage.