missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium cobalt(III) oxide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01404930
Description
Lithium cobalt(III) oxide is a critical substance in the quest for high performance, advanced batteries. It is used as a lithium ion cathode material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium cobalt(III) oxide | |
12190-79-3 | |
250 g | |
CoLiO2 | |
lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate | |
BFZPBUKRYWOWDV-UHFFFAOYSA-N | |
lithium;oxido(oxo)cobalt | |
23670860 | |
Odorless | |
Dark blue powder |
Blue | |
Powder | |
97% | |
MFCD00049786 | |
Insoluble in water. | |
[Li+].[O-][Co]=O | |
97.871 | |
97.88 | |
Hygroscopic |
Chemical Identifiers
12190-79-3 | |
97.871 | |
BFZPBUKRYWOWDV-UHFFFAOYSA-N | |
23670860 | |
[Li+].[O-][Co]=O |
CoLiO2 | |
MFCD00049786 | |
lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate | |
lithium;oxido(oxo)cobalt |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H350-H361f
EINECSNumber : 235-362-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only