missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium chloride, ultra dry, 99.995% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4476214

 View more versions of this product

Catalog No. AA4476214



Specifications

Specifications

Lithium chloride, ultra dry
99.995% (Metal basis)
MFCD00011078
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
[Li+].[Cl-]
42.39
CHEBI:48607
Crystalline Powder
White to Gray
7447-41-8
ClLi
White to gray crystalline powder
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium;chloride
433294
42.39
Odorless
25g

Chemical Identifiers

7447-41-8
42.39
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
433294
lithium;chloride
ClLi
MFCD00011078
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
CHEBI:48607
[Li+].[Cl-]
Safety and Handling

Safety and Handling

H302-H315-H319-H335: Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 231-212-3

SDS Documents