missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium chloride, ultra dry, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1454009

 View more versions of this product

Catalog No. AA1454009



Description

Description

  • Ampouled under argon
Specifications

Specifications

Lithium chloride
7447-41-8
ClLi
14,5528
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium;chloride
433294
42.39
99.9%
−10 Mesh
Ampule
1325°C to 1360°C
10g
Ultra dry
(metals basis)
MFCD00011078
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
[Li+].[Cl-]
42.39
CHEBI:48607
Beads
2.068g/mL
Odorless
Hygroscopic
605°C

Chemical Identifiers

7447-41-8
42.39
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
433294
lithium;chloride
ClLi
MFCD00011078
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
CHEBI:48607
[Li+].[Cl-]
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : OJ5950000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Documents