Please login to your online account to display your discounted pricing

Lithium Chloride, Molecular Biology Grade, Calbiochem™, EMD Millipore

A direct and noncompetitive inhibitor of GSK-3 (Ki = 2 mM) that activates Wnt-signaling

$46.27 - $69.77

Chemical Identifiers

Assay Percent Range ≥99%
Formula Weight 42.4
CAS 7447-41-8
Chemical Name or Material Lithium Chloride
Molecular Formula LiCl
Purity Grade Notes by argentometry
Solubility Information Easily soluble in cold water
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

438002250GM

 
emd millipore corporation
438002250GM
Each for $69.77

43800250GM

 
emd millipore corporation
43800250GM
Each for $46.27
Description & Specifications

Specifications

Formula Weight 42.4
Quantity 250g
Molecular Formula LiCl
Assay Percent Range ≥99%
RNase None detected
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Purity Grade Notes by argentometry
CAS 7447-41-8
RTECS Number OJ5950000
DNase None detected
Solubility Information Easily soluble in cold water
Chemical Name or Material Lithium Chloride
Safety and Handling

Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).