missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium chloride hydrate, White granules, 99.996% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1073522

 View more versions of this product

Catalog No. AA1073522



Specifications

Specifications

Lithium chloride hydrate
99.996% (Metals basis)
MFCD00149764
VXJIMUZIBHBWBV-UHFFFAOYSA-M
lithium;chloride;hydrate
23681138
Granules
White
85144-11-2
ClH2LiO
acmc-1bre5
[Li+].O.[Cl-]
60.405
42.39 (Anhydrous)
Odorless
100g

Chemical Identifiers

85144-11-2
60.405
VXJIMUZIBHBWBV-UHFFFAOYSA-M
23681138
[Li+].O.[Cl-]
ClH2LiO
MFCD00149764
acmc-1bre5
lithium;chloride;hydrate
Safety and Handling

Safety and Handling

H301-H315-H319-H335: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 231-212-3

Documents