Please login to your online account to display your discounted pricing

Lithium Chloride ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Lithium Chloride
CAS 7447-41-8
Purity Grade ACS Grade
Physical Form Granular
Molecular Formula LiCl
Formula Weight 42.39
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK5852125

 
avantor performance
585202
POLYSTORMOR 125g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK5852500

 
avantor performance
585204
POLYSTORMOR 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK5852212

  SDS
avantor performance
585206
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 42.39
Chemical Name or Material Lithium Chloride
Molecular Formula LiCl
Physical Form Granular
Packaging POLYSTORMOR
CAS 7447-41-8
Quantity 125g