missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium chloride, 99%, for molecular biology, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC449041000

 View more versions of this product

Catalog No. AC449041000



Description

Description

Specifications

Specifications

Lithium chloride
98.5% min. (Argentometry)
LiCl
chlorku litu, chlorku litu polish, chlorolithium, chlorure de lithium, licl, lithium chloride, lithium chloride licl, lithiumchlorid, lithiumchloride, luthium chloride
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium;chloride
433294
42.38
Molecular Biology
Plastic bottle
2.06
605.0°C
7447-41-8
ClLi
15,5584
Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols,ether,pyridine,,nitrobenzene and acetone
[Li+].[Cl-]
42.38
CHEBI:48607
99%
2.0600g/mL
Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone
1382.0°C
100g

Chemical Identifiers

7447-41-8
42.38
chlorku litu, chlorku litu polish, chlorolithium, chlorure de lithium, licl, lithium chloride, lithium chloride licl, lithiumchlorid, lithiumchloride, luthium chloride
CHEBI:48607
[Li+].[Cl-]
ClLi
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
433294
lithium;chloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 231-212-3

SDS Documents