missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium chloride, 1M aq. soln., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J61895AE

 View more versions of this product

Catalog No. AAJ61895AE



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lithium chloride
7447-41-8
100 mL
LiCl
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
[Li+].[Cl-]
42.39
CHEBI:48607
1M aq. soln.
Colorless
Liquid
ClLi
MFCD00011078
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium(1+) chloride
433294
42.39
Odorless

Chemical Identifiers

7447-41-8
42.39
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
433294
[Li+].[Cl-]
ClLi
MFCD00011078
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
CHEBI:48607
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only