missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium Carbonate, puriss p.a, ≥99.0% (T), For Microscopy, Honeywell Fluka™
Puriss. p.a., ACS Reagent, reagent (for microscopy),≥99.0% (T)
$196.85 - $196.85
Chemical Identifiers
CAS | 554-13-2 |
---|---|
Molecular Formula | CLi2O3 |
Molecular Weight (g/mol) | 73.89 |
MDL Number | MFCD00011084 |
InChI Key | XGZVUEUWXADBQD-UHFFFAOYSA-L |
Synonym | lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine |
PubChem CID | 11125 |
ChEBI | CHEBI:6504 |
IUPAC Name | dilithium;carbonate |
SMILES | [Li+].[Li+].[O-]C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
60047222
|
Honeywell Chemicals
62470100G |
100 g | Poly bottle |
Each for $196.85
|
|
||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
554-13-2 | |
73.89 | |
XGZVUEUWXADBQD-UHFFFAOYSA-L | |
11125 | |
dilithium;carbonate |
CLi2O3 | |
MFCD00011084 | |
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
CHEBI:6504 | |
[Li+].[Li+].[O-]C([O-])=O |
Specifications
1265°C (1013hPa) | |
554-13-2 | |
Solid | |
CLi2O3 | |
MFCD00011084 | |
3999191 | |
XGZVUEUWXADBQD-UHFFFAOYSA-L | |
dilithium;carbonate | |
11125 | |
73.89g/mol | |
Poly bottle | |
Lithium carbonate |
550°C | |
Colorless | |
100 g | |
Li2CO3 | |
NONH for all modes of transport | |
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
[Li+].[Li+].[O-]C([O-])=O | |
73.89 | |
CHEBI:6504 | |
≥99.0% (T) | |
3.46g/cm3 |
Safety and Handling
P305 + P351 + P338
RTECSNumber : 209-062-5