Please login to your online account to display your discounted pricing

Lithium Carbonate ACS AR Powder, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Lithium Carbonate
CAS 554-13-2
Purity Grade ACS Grade
Physical Form Powder
Molecular Formula Li2CO3
Formula Weight 73.89
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK5840125

  SDS
avantor performance
584002
POLYSTORMOR 125g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 73.89
Physical Form Powder
Chemical Name or Material Lithium Carbonate
Packaging POLYSTORMOR
Molecular Formula Li2CO3
CAS 554-13-2
Quantity 125g