missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium carbonate, 99%, pure, ACROS Organics™

Manufacturer:  Acros Organics AC197780025

 View more versions of this product

Catalog No. AC197780025



Specifications

Specifications

Lithium carbonate
554-13-2
CLi2O3
MFCD00011084
carbonic acid lithium salt
[Li+].[Li+].C(=O)([O-])[O-]
73.888
CHEBI:6504
Finely Granulated Powder
Pure
0.4% max. (200°C)
Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids, practically insoluble in alcohol
720.0°C
99%
99%
Li2CO3
01,606; 03,183; 04,192; 05,396
XGZVUEUWXADBQD-UHFFFAOYSA-L
dilithium;carbonate
11125
73.88
99%
2.11
Plastic drum
White
2.5kg

Chemical Identifiers

554-13-2
73.888
XGZVUEUWXADBQD-UHFFFAOYSA-L
11125
dilithium;carbonate
CLi2O3
MFCD00011084
carbonic acid lithium salt
CHEBI:6504
[Li+].[Li+].C(=O)([O-])[O-]
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye damage.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 209-062-5

RTECSNumber : OJ5800000

TSCA : TSCA

SDS Documents