Please login to your online account to display your discounted pricing

Lithium carbonate, 99%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Lithium carbonate
CAS 554-13-2
Molecular Formula Li2CO3
Formula Weight 73.9
MDL Number MFCD00011084
Merck Index 14,5527
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA13418A3

  SDS
alfa aesar
13418A3
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 73.9
Quantity 2kg
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a;
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White
Assay 99%
MDL Number MFCD00011084
pH 10 to 11.5
Solubility Information Soluble in water and dilute acids. Insoluble in alcohol
Odor Odorless
Health Hazard 2 GHS H Statement
H302-H319;
Harmful if swallowed.
Causes serious eye irritation.
Boiling Point 1310°C (dec.)
Health Hazard 1 Warning
CAS 554-13-2
Molecular Formula Li2CO3
Merck Index 14,5527
Melting Point 723°C
RTECS Number OJ5800000
Chemical Name or Material Lithium carbonate
Density 2.11
Physical Form Powder
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a;
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H319;
Harmful if swallowed.
Causes serious eye irritation.