missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium carbonate, 99.99% (metals basis excluding Ca), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 10734A1

 View more versions of this product

Catalog No. AA10734A1



Specifications

Specifications

Lithium carbonate
99.998% (Metals basis excluding Ca)
MFCD00011084
carbonic acid lithium salt
[Li+].[Li+].C(=O)([O-])[O-]
73.888
CHEBI:6504
Powder
White
554-13-2
CLi2O3
White powder
XGZVUEUWXADBQD-UHFFFAOYSA-L
dilithium;carbonate
11125
73.9
Odorless
1kg

Chemical Identifiers

554-13-2
73.888
XGZVUEUWXADBQD-UHFFFAOYSA-L
11125
dilithium;carbonate
CLi2O3
MFCD00011084
carbonic acid lithium salt
CHEBI:6504
[Li+].[Li+].C(=O)([O-])[O-]
Safety and Handling

Safety and Handling

H318-H315-H302-H335: Causes serious eye damage. Causes skin irritation. Harmful if swallowed. May cause respiratory irritation.

EINECSNumber : 209-062-5

Documents