Learn More
Lithium carbonate, Puratronic™, 99.998% (metals basis excluding Ca), Ca LT 20ppm, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01073418
Description
Lithium carbonate-based glasses are used in overware. Used in the manufacture of lithium-ion battery cathodes. Pharmaceutical composition of lithium carbonate has been shown to exhibit antimanic effects specifically to treat mania associated with bipolar disorder. Used in both low-fire and high-fire ceramic glaze, tile adhesives, and in the processing of metal oxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1310°C (decomposition) | |
723°C | |
White | |
50 g | |
CLi2O3 | |
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
XGZVUEUWXADBQD-UHFFFAOYSA-L | |
dilithium(1+) carbonate | |
11125 | |
73.9 | |
White powder |
Lithium carbonate | |
554-13-2 | |
Powder | |
99.998% (Metals basis excluding Ca) | |
MFCD00011084 | |
Soluble in water and acid. Insoluble in acetone,ammonia and alcohol. | |
[Li+].[Li+].[O-]C([O-])=O | |
73.89 | |
CHEBI:6504 | |
Odorless | |
2.11 g/cm3 at 20°C |
Chemical Identifiers
554-13-2 | |
73.89 | |
XGZVUEUWXADBQD-UHFFFAOYSA-L | |
11125 | |
[Li+].[Li+].[O-]C([O-])=O |
CLi2O3 | |
MFCD00011084 | |
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
CHEBI:6504 |
Safety and Handling
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
EINECSNumber : 209-062-5
RTECSNumber : OJ5800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only