missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium bromide, White beads, 99.998% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1453706

 View more versions of this product

Catalog No. AA1453706



Specifications

Specifications

Lithium bromide
99.998% (Metals basis)
MFCD00011077
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium;bromide
82050
86.85
White
7550-35-8
BrLi
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
[Li+].[Br-]
86.844
CHEBI:63042
Beads
5g

Chemical Identifiers

7550-35-8
86.844
AMXOYNBUYSYVKV-UHFFFAOYSA-M
82050
lithium;bromide
BrLi
MFCD00011077
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
CHEBI:63042
[Li+].[Br-]
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.

H302-H315-H319-H317: Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.

EINECSNumber : 231-439-8

Documents