missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium bromide, ultra dry, White powder, 99.998% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4419914

 View more versions of this product

Catalog No. AA4419914



Specifications

Specifications

Lithium bromide, ultra dry
99.998% (Metals basis)
MFCD00011077
Soluble in water (at 20°C 1490g/L),alcohol and glycol.
[Li+].[Br-]
86.844
CHEBI:63042
Powder
Hygroscopic
White
25g
7550-35-8
BrLi
bromolithium
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium;bromide
82050
86.85
3.464
1265°C
550°C

Chemical Identifiers

7550-35-8
86.844
AMXOYNBUYSYVKV-UHFFFAOYSA-M
82050
lithium;bromide
BrLi
MFCD00011077
bromolithium
CHEBI:63042
[Li+].[Br-]
Safety and Handling

Safety and Handling

P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c

H302-H315-H317-H319

EINECSNumber : 231-439-8

RTECSNumber : OJ5755000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents