missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium bromide, ultra dry, White powder, 99.998% (Metals basis), Alfa Aesar™

Chemical Identifiers

CAS 7550-35-8
Physical Form Powder
Assay Percent Range 99.998% (Metals basis)
Molecular Formula BrLi
Formula Weight 86.85
MDL Number MFCD00011077
Chemical Name or Material Lithium bromide, ultra dry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4419906 SDS Alfa Aesar
4419906
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4419914 SDS Alfa Aesar
4419914
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4419922 SDS Alfa Aesar
4419922
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7550-35-8
99.998% (Metals basis)
86.85
Lithium bromide, ultra dry
Powder
BrLi
MFCD00011077

Specifications

Powder
5g
BrLi
99.998% (Metals basis)
Lithium bromide, ultra dry
86.85
White
7550-35-8
MFCD00011077

Safety and Handling

H302-H315-H319-H317: Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.

EINECSNumber : 231-439-8