Please login to your online account to display your discounted pricing

Lithium bromide, ultra dry, 99.9% (metals basis), Alfa Aesar™

$128.38 - $128.38

Chemical Identifiers

Chemical Name or Material Lithium bromide, ultra dry
CAS 7550-35-8
Physical Form Beads
Assay Percent Range 99.9% (Metal basis)
Molecular Formula BrLi
Formula Weight 86.85
MDL Number MFCD00011077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3570514

  SDS
alfa aesar
3570514
25g Each for $128.38

AA3570522

  SDS
alfa aesar
3570522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 86.85
Quantity 25g
Color White
Density 3.464g/cm3 at 20°C
Physical Form Beads
EINECS Number 231-439-8
Chemical Name or Material Lithium bromide, ultra dry
MDL Number MFCD00011077
Health Hazard 1 H302-H315-H319-H317: Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.
Assay Percent Range 99.9% (Metal basis)
Appearance White beads
CAS 7550-35-8
Molecular Formula BrLi
Safety and Handling

H302-H315-H319-H317: Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.