missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Bromide Solution, Ultra-Pure Additive, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Manufacturer:  Spex Certiprep Llc FFB10303

 View more versions of this product

Catalog No. FFB10303



Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Ultra Pure Additives
  • Lithium Bromide Solution
  • 15 mL Package Size

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Lithium Bromide Solution
Additive Ultra Pure
BrLi
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium;bromide
82050
Ultrapure
15mL
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
7550-35-8
bromolithium, libr, lithium 1+ ion bromide, lithium bromide, lithium bromide libr, lithium bromide, anhydrous, lithium bromide, ultra dry, lithium monobromide, lithium-bromide, lithiumbromide
[Li+].[Br-]
86.844
CHEBI:63042
Triple-leached LDPE bottle
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Chemical Identifiers

7550-35-8
86.844
bromolithium, libr, lithium 1+ ion bromide, lithium bromide, lithium bromide libr, lithium bromide, anhydrous, lithium bromide, ultra dry, lithium monobromide, lithium-bromide, lithiumbromide
CHEBI:63042
[Li+].[Br-]
BrLi
AMXOYNBUYSYVKV-UHFFFAOYSA-M
82050
lithium;bromide
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Product intended for laboratory use only