Learn More
Lithium bromide, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 199871000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1265.0°C | |
550.0°C | |
7550-35-8 | |
6 to 12 (4% soln. at 20°C) | |
100 g | |
BrLi | |
MFCD00011077 | |
15,5582 | |
Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol, ether, methanol,, acetone and glycol, slightly soluble in pyridine, soluble in amyl alcohol | |
[Li+].[Br-] | |
86.84 | |
CHEBI:63042 | |
≥99% | |
0.5% max. (400°C) | |
3.46 |
Lithium bromide | |
99+% | |
Gray to Black | |
Powder | |
99% min. (Argentometry) | |
LiBr | |
01,604; 02,245; 04,297 | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
lithium;bromide | |
82050 | |
86.84 | |
Analytical | |
Plastic bottle | |
3.4600g/mL |
Chemical Identifiers
7550-35-8 | |
86.84 | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
82050 | |
lithium;bromide |
BrLi | |
MFCD00011077 | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
CHEBI:63042 | |
[Li+].[Br-] |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protect
GHS Signal Word: Warning
EINECSNumber : 231-439-8
RTECSNumber : OJ5755000
TSCA : TSCA
RUO – Research Use Only