Learn More
Lithium bromide, 99%, pure, anhydrous, Thermo Scientific Chemicals
$402.36 - $774.51
Chemical Identifiers
CAS | 7550-35-8 |
---|---|
Molecular Formula | BrLi |
Molecular Weight (g/mol) | 86.84 |
InChI Key | AMXOYNBUYSYVKV-UHFFFAOYSA-M |
Synonym | lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide |
PubChem CID | 82050 |
ChEBI | CHEBI:63042 |
IUPAC Name | lithium;bromide |
SMILES | [Li+].[Br-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC453980010
|
Thermo Scientific Chemicals
453980010 |
1 kg | Plastic Bottle |
Each for $402.36
|
|
||||
AC453980025
|
Thermo Scientific Chemicals
453980025 |
2.5 kg | Plastic Bottle |
Each for $774.51
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7550-35-8 | |
86.84 | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
CHEBI:63042 | |
[Li+].[Br-] |
BrLi | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
82050 | |
lithium;bromide |
Specifications
1265.0°C | |
7550-35-8 | |
1 kg | |
BrLi | |
15,5582 | |
Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol, ether, methanol,, acetone and glycol, slightly soluble in pyridine, soluble in amyl alcohol | |
[Li+].[Br-] | |
86.84 | |
CHEBI:63042 | |
99% | |
Plastic Bottle | |
3.4600g/mL |
550.0°C | |
6 to 12 (4 % aq. soln.) | |
98.5% min. (Argentometry) | |
LiBr | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
lithium;bromide | |
82050 | |
86.84 | |
Pure | |
3.46 | |
Lithium Bromide |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protect
GHS Signal Word: Warning
EINECSNumber : 231-439-8
RUO – Research Use Only