Learn More
Lithium bromide, 99.999%, (trace metal basis), extra pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 212532500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For ultra trace analysisSpecifications
Lithium bromide | |
7550-35-8 | |
99.999% | |
LiBr | |
01,604; 02,245; 04,297 | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
lithium;bromide | |
82050 | |
86.84 | |
Trace Metal Basis | |
3.46 |
Extra Pure | |
250g | |
BrLi | |
MFCD00011077 | |
15, 5582 | |
Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol,ether,methanol,,acetone and glycol,slightly soluble in pyridine,soluble in amyl alcohol | |
[Li+].[Br-] | |
86.84 | |
CHEBI:63042 | |
99.999% | |
Glass bottle | |
3.4600g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protect
GHS Signal Word: Warning
EINECSNumber : 231-439-8
RUO â Research Use Only