Please login to your online account to display your discounted pricing

Lithium bromide, 4M solution in THF, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC390971000

 View more versions of this product

Catalog No. AC390971000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 86.84
Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 1.1800g/mL
Merck Index 15, 5582
Chemical Name or Material Lithium bromide
Molecular Formula BrLi
Linear Formula LiBr
Appearance of Solution Clear colorless to yellowish solution
MDL Number MFCD00011077
CAS 7550-35-8
Health Hazard 1 Danger
CAS 109-99-9
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye
Concentration 4M exact strength on the certificate of analysis
Health Hazard 2 GHS P Statement Harmful if swallowed. Causes serious eye irritation. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides.

Specifications

Chemical Name or Material Lithium bromide
CAS 7550-35-8
CAS 109-99-9
Molecular Formula BrLi
Linear Formula LiBr
Formula Weight 86.84
MDL Number MFCD00011077
Accessories

Danger

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye

GHS P Statement Harmful if swallowed. Causes serious eye irritation. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides.

SDS & Safety Documents