Please login to your online account to display your discounted pricing

Lithium bromide, 4M solution in THF, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Lithium bromide
CAS 7550-35-8
CAS 109-99-9
Molecular Formula BrLi
Linear Formula LiBr
Formula Weight 86.84
MDL Number MFCD00011077
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390971000

  SDS
thermo fisher scientific
AC390971000
AcroSeal™ Glass Bottle in Tin Can 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC390978000

  SDS
thermo fisher scientific
AC390978000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 2 GHS H Statement
May cause drowsiness or dizziness. Causes serious eye irritation. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. Causes skin irritation. May cause an allergic skin reaction. Suspected of causing cancer.
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wash face, hands and any exposed skin thoroughly after handling. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. If skin irritation occurs: Get medical advice/attention.
Formula Weight 86.84
Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 1.1800g/mL
Merck Index 15, 5582
Chemical Name or Material Lithium bromide
CAS 109-99-9
Appearance of Solution Clear colorless to yellowish solution
Linear Formula LiBr
Concentration 4M exact strength on the certificate of analysis
Molecular Formula BrLi
CAS 7550-35-8
MDL Number MFCD00011077
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wash face, hands and any exposed skin thoroughly after handling. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. If skin irritation occurs: Get medical advice/attention.

GHS H Statement
May cause drowsiness or dizziness. Causes serious eye irritation. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. Causes skin irritation. May cause an allergic skin reaction. Suspected of causing cancer.