missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium borohydride, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 206810500

 View more versions of this product

Catalog No. AC206810500



Description

Description

Reduction reagent
Specifications

Specifications

Lithium borohydride
16949-15-8
H4BLi
MFCD00011088
15, 5581
Solubility in water: 209g/L (10°C). Other solubilities: soluble in ether,ethanol,tetrahydrofuran and,aliphatic amines
[Li+].[B-]
21.77
21.77
95%
Glass Bottle
0.66
280.0°C
95%
95%
LiBH4
01,603; 04,296; 11,293; 12,276; 13,92; 14,191; 15,186
bh4li, borate 1-, tetrahydro-, lithium, borate 1-, tetrahydro-, lithium 1:1, boryllithium, lithium 1+ ion boranuide, lithium borohydride 2.0 m in thf, lithium hydroborate, lithium, boryl, lithotab borohydride, unii-8l87x4s4kp
NZTNZPDOBQDOSO-UHFFFAOYSA-N
lithium;boron(1-)
20722760
Powder
0.6600g/mL
Solubility in water: 209g/L (10°C). Other solubilities: soluble in ether, ethanol, tetrahydrofuran and, aliphatic amines
White
50g

Chemical Identifiers

16949-15-8
21.77
NZTNZPDOBQDOSO-UHFFFAOYSA-N
20722760
[Li+].[B-]
H4BLi
MFCD00011088
bh4li, borate 1-, tetrahydro-, lithium, borate 1-, tetrahydro-, lithium 1:1, boryllithium, lithium 1+ ion boranuide, lithium borohydride 2.0 m in thf, lithium hydroborate, lithium, boryl, lithotab borohydride, unii-8l87x4s4kp
lithium;boron(1-)
Safety and Handling

Safety and Handling

GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Reacts violentl

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face p

GHS Signal Word: Danger

EINECSNumber : 241-021-7

RTECSNumber : ED2725000

TSCA : TSCA

SDS Documents