missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium bis(trimethylsilyl)amide, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC338142500

 View more versions of this product

Catalog No. AC338142500



Description

Description

Specifications

Specifications

Lithium bis(trimethylsilyl)amide
Authentic
0.891
Yellow
4039-32-1
C6H18LiNSi2
MFCD00008261
hexamethyldisilazane lithium salt, lhmds, lihmds, lithium bis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium bis-trimethylsilyl amide, lithium hexamethyldisilazane, lithium hexamethyldisilazide, lithiumbis trimethylsilyl amide, unii-rc4n1i108m
YNESATAKKCNGOF-UHFFFAOYSA-N
lithium;bis(trimethylsilyl)azanide
2733832
Crystalline Powder or Chunks
0.8910g/mL
Glass bottle
114°C to 116°C
250g
95%
[(CH3)3Si]2NLi
04,296; 05,393; 07,197; 12,280; 13,165; 14,194
Solubility in water: decomposes
[Li+].C[Si](C)(C)[N-][Si](C)(C)C
167.33
167.33
95%

Chemical Identifiers

4039-32-1
167.33
YNESATAKKCNGOF-UHFFFAOYSA-N
2733832
[Li+].C[Si](C)(C)[N-][Si](C)(C)C
C6H18LiNSi2
MFCD00008261
hexamethyldisilazane lithium salt, lhmds, lihmds, lithium bis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium bis-trimethylsilyl amide, lithium hexamethyldisilazane, lithium hexamethyldisilazide, lithiumbis trimethylsilyl amide, unii-rc4n1i108m
lithium;bis(trimethylsilyl)azanide
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse

GHS Signal Word: Danger

EINECSNumber : 223-725-6

SDS Documents