missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium bis(trimethylsilyl)amide, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC338140500
Specifications
Lithium bis(trimethylsilyl)amide | |
Authentic | |
0.891 | |
Yellow | |
4039-32-1 | |
C6H18LiNSi2 | |
MFCD00008261 | |
hexamethyldisilazane lithium salt, lhmds, lihmds, lithium bis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium bis-trimethylsilyl amide, lithium hexamethyldisilazane, lithium hexamethyldisilazide, lithiumbis trimethylsilyl amide, unii-rc4n1i108m | |
YNESATAKKCNGOF-UHFFFAOYSA-N | |
lithium;bis(trimethylsilyl)azanide | |
2733832 | |
Crystalline Powder or Chunks |
0.8910g/mL | |
Glass bottle | |
114°C to 116°C | |
50g | |
95% | |
[(CH3)3Si]2NLi | |
04,296; 05,393; 07,197; 12,280; 13,165; 14,194 | |
Solubility in water: decomposes | |
[Li+].C[Si](C)(C)[N-][Si](C)(C)C | |
167.33 | |
167.33 | |
95% |
Chemical Identifiers
4039-32-1 | |
167.33 | |
YNESATAKKCNGOF-UHFFFAOYSA-N | |
2733832 | |
[Li+].C[Si](C)(C)[N-][Si](C)(C)C |
C6H18LiNSi2 | |
MFCD00008261 | |
hexamethyldisilazane lithium salt, lhmds, lihmds, lithium bis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium bis-trimethylsilyl amide, lithium hexamethyldisilazane, lithium hexamethyldisilazide, lithiumbis trimethylsilyl amide, unii-rc4n1i108m | |
lithium;bis(trimethylsilyl)azanide |
Safety and Handling
GHS H Statement
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
GHS Signal Word: Danger
EINECSNumber : 223-725-6